Boron nitride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
255475-250G 255475-50G
SIAL255475-250GEA 608 EUR
SIAL255475-250G SIAL255475-50G
Boron nitride, Sigma-Aldrich®
Boron nitride
Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527
Density: 2.15 g/cm³ (20 °C)
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336