Triisopropyl borate 98+%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Tri-i-propylborate
188740800 188747500
ACRO188740800EA 56.3 EUR
ACRO188740800 ACRO188747500
Triisopropyl borate 98+%
Triisopropyl borate
Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139
Melting Pt: –59 °C
Density: 0.814 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3755 to 1.3775 (20°C, 589 nm)
GC ≥98.0 %