Tributyl borate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tri-n-butylborate
90795-100ML 90795-500ML
SIAL90795-100MLEA 97.2 EUR
SIAL90795-100ML SIAL90795-500ML
Tributyl borate, Sigma-Aldrich®
Tributyl borate
Formula: C₁₂H₂₇BO₃
MW: 230.15 g/mol
Boiling Pt: 230
Melting Pt: –70 °C
Density: 0.8567 g/cm³ (20 °C)
MDL Number: MFCD00009434
CAS Number: 688-74-4
EINECS: 211-706-5