Triethyl borate, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

T59307-100ML T59307-500ML
SIALT59307-100MLEA 66.4 EUR
SIALT59307-100ML SIALT59307-500ML
Triethyl borate, Sigma-Aldrich®
Triethyl borate
Formula: C₆H₁₅BO₃
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –84.8 °C
Density: 0.858 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR