Orthoboric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
MQ200
B6768-10KG B6768-1KG B6768-500G B6768-5KG
SIALB6768-10KGEA 562 EUR
SIALB6768-10KG SIALB6768-1KG SIALB6768-500G SIALB6768-5KG
Orthoboric acid, Sigma-Aldrich®
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM