Thallium(I) sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Thallic sulphate, Dithallium sulphate
M-Clarity™ quality level = MQ100
204625-25G 204625-5G
SIAL204625-25GEA 196 EUR
SIAL204625-25G SIAL204625-5G
Thallium(I) sulphate, Sigma-Aldrich®
Dithallium sulphate
Formula: O₄STl₂
MW: 504.83 g/mol
Melting Pt: 632 °C
Density: 6.77 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011278
CAS Number: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II
Merck Index: 12,09405