Diisobutylaluminium hydride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL256811-100GEA 107 EUR
SIAL256811-100G
Diisobutylaluminium hydride, Sigma-Aldrich®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142,22 g/mol
Boiling Pt: 95,5…96.5 °C (0.2 mmHg)
Melting Pt: –80 °C
Density: 0,798 g/cm³ (20 °C)
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR