Silicon carbide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
378097-1KG 378097-250G
SIAL378097-1KGEA 176 EUR
SIAL378097-1KG SIAL378097-250G
Silicon carbide, Sigma-Aldrich®
Silicon carbide
Formula: SiC
MW: 40.1 g/mol
Melting Pt: 2700
Density: 3.22 g/cm³ (25 °C)
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM