Lead(II) chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL449865-5GEA 219 EUR
SIAL449865-5G
Lead(II) chloride, Sigma-Aldrich®
Lead(II) chloride
Formula: Cl₂Pb
MW: 278,11 g/mol
Boiling Pt: 950 °C (1013 hPa)
Melting Pt: 496 °C
Density: 5,85 g/cm³ (20 °C)
MDL Number: MFCD00011157
UN: 2291
ADR: 6.1,III
Merck Index: 13,05422