Lead(II) nitrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Lead dinitrate
M-Clarity™ quality level = MQ100
467790-250G 467790-50G
SIAL467790-250GEA 436 EUR
SIAL467790-250G SIAL467790-50G
Lead(II) nitrate, Sigma-Aldrich®
Lead (II) nitrate
Formula: Pb(NO₃)₂
Melting Pt: 450…470 °C
Density: 4.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432