Tin(IV) chloride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

217913-100G 217913-25G 217913-5G
SIAL217913-100GEA 322 EUR
SIAL217913-100G SIAL217913-25G SIAL217913-5G
Tin(IV) chloride, Sigma-Aldrich®
Tin (IV) chloride
Formula: Cl₄Sn
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –33 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 12,08929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR