Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Supplier: Thermo Fisher Scientific
Warning
190000100 190000500 190002500
ACRO190000100EA 121 EUR
ACRO190000100 ACRO190000500 ACRO190002500
Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Germanium (IV) oxide
Formula: GeO₂
Melting Pt: 1115 °C
Density: 3,64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤10 ppm