Germanium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Germanium dioxide
MQ200
199478-100G 199478-10G 199478-25G
SIAL199478-100GEA 866 EUR
SIAL199478-100G SIAL199478-10G SIAL199478-25G
Germanium(IV) oxide, Sigma-Aldrich®
Germanium (IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1110
Density: 3.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417