Diatomaceous earth (natural) filter aid

Supplier: Roth Carl

8019.1 8019.2
ROTH8019.1EA 61.7 EUR
ROTH8019.1 ROTH8019.2
Diatomaceous earth (natural) filter aid
Diatomaceous earth
Formula: SiO₂
MDL Number: MFCD00132803
CAS Number: 61790-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR