Silicon dioxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
M-Clarity™ quality level = MQ100
637246-250G 637246-500G 637246-50G
SIAL637246-250GEA 678 EUR
SIAL637246-250G SIAL637246-500G SIAL637246-50G
Silicon dioxide, Sigma-Aldrich®
Silicium dioxide
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1719
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493