Lead(II) sulphide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Lead sulphide
M-Clarity™ quality level = MQ100
372595-100G 372595-25G
SIAL372595-100GEA 333 EUR
SIAL372595-100G SIAL372595-25G
Lead(II) sulphide, Sigma-Aldrich®
Lead (II) sulphide
Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344
Melting Pt: 1114
Density: 7.5 g/cm³ (20 °C)
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9 III
Merck Index: 12,05445