Tin(II) sulphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tin(II) sulphate, Tin sulphate, Stannous sulphate
M-Clarity™ quality level = MQ100
244635-100G 244635-500G
SIAL244635-100GEA 50.2 EUR
SIAL244635-100G SIAL244635-500G
Tin(II) sulphate, Sigma-Aldrich®
Tin (II) sulphate
Formula: O₄SSn
MW: 214.77 g/mol
Density: 4.15 g/cm³ (20 °C)
MDL Number: MFCD00011246
CAS Number: 7488-55-3
EINECS: 231-302-2
Merck Index: 12,08947