Vinyltriethoxysilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Triethoxy(vinyl)silane
M-Clarity™ quality level = MQ200
175560-100ML 175560-500ML
SIAL175560-100MLEA 62.3 EUR
SIAL175560-100ML SIAL175560-500ML
Vinyltriethoxysilane, Sigma-Aldrich®
Vinyltriethoxysilane
Formula: C₈H₁₈O₃Si
MW: 190.31 g/mol
Boiling Pt: 160 °C (1013 hPa)
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00009063
CAS Number: 78-08-0
EINECS: 201-081-7