(3-Glycidoxypropyl)trimethoxysilane 97%
Supplier: Thermo Scientific
Danger
Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane
216540050 216541000 216545000 216540025
ACRO216540050EA 28.5 EUR
ACRO216540050 ACRO216541000 ACRO216545000 ACRO216540025
(3-Glycidoxypropyl)trimethoxysilane 97%
(3-Glycidoxypropyl)trimethoxysilane
Formula: C₉H₂₀O₅Si
MW: 236.34 g/mol
Boiling Pt: 120 °C (2 mmHg)
Melting Pt: 0 °C
Density: 1.07 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤50 APHA