Butyltin trichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: n-Butyltin trichloride, Trichlorobutylstannane

M-Clarity™ quality level = MQ100

201057-500G 201057-100G 201057-5G
SIAL201057-500GEA 214 EUR
SIAL201057-500G SIAL201057-100G SIAL201057-5G
Butyltin trichloride, Sigma-Aldrich®
Butyltin trichloride
Formula: C₄H₉Cl₃Sn
MW: 282.18 g/mol
Boiling Pt: 227 °C (1013 hPa)
Density: 1.71 g/cm³ (25 °C)
MDL Number: MFCD00000515
CAS Number: 1118-46-3
EINECS: 214-263-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR