(3-Aminopropyl)triethoxysilane
Supplier: Molekula
Danger
Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane
15593108-500G 15593108-100G
MOLE15593108-500GEA 140 EUR
MOLE15593108-500G MOLE15593108-100G
(3-Aminopropyl)triethoxysilane
(3-Aminopropyl)triethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: < –70 °C
Density: 0,95 g/cm³ (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4