(3-Aminopropyl)triethoxysilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane
M-Clarity™ quality level = MQ200
440140-100ML 440140-500ML
SIAL440140-100MLEA 108 EUR
SIAL440140-100ML SIAL440140-500ML
(3-Aminopropyl)triethoxysilane, Sigma-Aldrich®
(3-Aminopropyl)triethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217
Melting Pt: < –70 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 104
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4