(3-Mercaptopropyl)trimethoxysilane 95%
Supplier: Thermo Scientific
Warning
Synonyms: Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol
B23726.14 B23726.22 B23726.36
B23726.14EA 35.7 EUR
B23726.14 B23726.22 B23726.36
(3-Mercaptopropyl)trimethoxysilane 95%
(3-Mercaptopropyl)trimethoxysilane
Purity: 95% CAS No.: 4420-74-0 Molecular Formula: C6H16O3SSi Formula Weight: 196.34 Flash Point: 96°(204°F) Refractive Index: 1.4430 Melting Point: -50° Boiling Point: 213-215° Beilstein Reference: 2038119 UN No.: 3082 MDL No.: MFCD00004901
Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
MW: 196.34 g/mol
Boiling Pt: 213…215 °C
Melting Pt: < –50 °C
Density: 1.050
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004901
CAS Number: 4420-74-0
EINECS: 224-588-5
UN: 3082
ADR: 9,III