Dibutilkositrov diklorid

Supplier: EHRENSTORFER GMBH

EHERC12258000EA 49.6 EUR
EHERC12258000
Dibutilkositrov diklorid
Dibutilkositrov diklorid
Formula: C₈H₁₈Cl₂Sn
MW: 303,85 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1,37…1.4 g/cm³ (20 °C)
MDL Number: MFCD00000518
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR