Dibutilkositrov diklorid
Supplier: LGC Standards PROMOCHEM
PROODE-MS-03438EA
197
EUR
PROODE-MS-03438
Dibutilkositrov diklorid
Dibutilkositrov diklorid
Formula:
C₈H₁₈Cl₂Sn MW: 303,85 g/mol Boiling Pt: 275 °C (1013 hPa) Melting Pt: 37…39 °C Density: 1,37…1.4 g/cm³ (20 °C) |
MDL Number:
MFCD00000518 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...