Dibutyltin dichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC
M-Clarity™ quality level = MQ100
205494-250G
205494-50G
SIAL205494-250GEA
96.3
EUR
SIAL205494-250G
SIAL205494-50G
Dibutyltin dichloride, Sigma-Aldrich®
Dibutyltin dichloride
Formula:
C₈H₁₈Cl₂Sn MW: 303.85 g/mol Boiling Pt: 275 °C (1013 hPa) Melting Pt: 37…39 °C Density: 1.37…1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000518 CAS Number: 683-18-1 EINECS: 211-670-0 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...