Dimethyldichlorosilane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Dichloro(dimethyl)silane, Dichlorodimethylsilane
40140-100ML
40140-25ML
40140-500ML
SIAL40140-100MLEA
54.8
EUR
SIAL40140-100ML
SIAL40140-25ML
SIAL40140-500ML
Dimethyldichlorosilane, Sigma-Aldrich®
Dimethyldichlorosilane
Formula:
C₂H₆Cl₂Si MW: 129.06 g/mol Boiling Pt: 70 °C (1013 hPa) Melting Pt: –76 °C Density: 1.0745 g/cm³ (20 °C) Flash Pt: –16 °C |
MDL Number:
MFCD00000491 CAS Number: 75-78-5 EINECS: 200-901-0 UN: 1162 ADR: 3,II REACH: 01-2119437250-51 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...