Dimethyltin dichloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Dichlorodimethylstannane

M-Clarity™ quality level = MQ100

288012-25G 288012-5G
SIAL288012-25GEA 436 EUR
SIAL288012-25G SIAL288012-5G
Dimethyltin dichloride, Sigma-Aldrich®
Dimethyltin dichloride
Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 124 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR