Bromotrimethylsilane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethylbromosilane, Trimethylsilyl bromide
M-Clarity™ quality level = MQ200
92337-100ML 92337-25ML 92337-5ML
SIAL92337-100MLEA 221 EUR
SIAL92337-100ML SIAL92337-25ML SIAL92337-5ML
Bromotrimethylsilane, Sigma-Aldrich®
Bromotrimethylsilane
Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: –43 °C
Density: 1.1895 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000048
CAS Number: 2857-97-8
EINECS: 220-672-0
UN: 2924
ADR: 3,II