Triisopropylchlorosilane ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 97+%
CAS No.: 13154-24-0
Molecular Formula: [(CH3)2CH]3SiCl
Formula Weight: 192.81
Flash Point: 62°(144°F)
Refractive Index: 1.4535
Odour: Acrid
Boiling Point: 200°
Beilstein Reference: 1737446
UN No.: 2987
MDL No.: MFCD00009656
Formula:
C₉H₂₁ClSi Boiling Pt: 200 °C Density: 0.908 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009656 CAS Number: 13154-24-0 UN: 2987 ADR: 8,II |