Chlorotriethylsilane 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Triethylchlorosilane
215940250 215941000 215945000
ACRO215940250EA 74.3 EUR
ACRO215940250 ACRO215941000 ACRO215945000
Chlorotriethylsilane 99%
Chlorotriethylsilane
Formula: C₆H₁₅ClSi
MW: 150.72 g/mol
Boiling Pt: 142…144 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥98.5 %