Phenyltriethoxysilane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phenyltriethoxysilane, Triethoxy(phenyl)silane, Triethoxyphenylsilane
M-Clarity™ quality level = MQ200
175609-1KG 175609-250G 175609-5G
SIAL175609-1KGEA 167 EUR
SIAL175609-1KG SIAL175609-250G SIAL175609-5G
Phenyltriethoxysilane, Sigma-Aldrich®
Phenyltriethoxysilane
Formula: C₁₂H₂₀O₃Si
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 41 °C
Density: 0.996 g/cm³ (20 °C)
MDL Number: MFCD00009065
CAS Number: 780-69-8
EINECS: 212-305-8