Phenylsilane ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Silylbenzene
A17901.06 A17901.14 A17901.22
A17901.06EA 81 EUR
A17901.06 A17901.14 A17901.22
Phenylsilane ≥97%
Phenylsilane
Purity: 97% CAS No.: 694-53-1 Molecular Formula: C6H8Si Formula Weight: 108.22 Flash Point: 7°(45°F) Refractive Index: 1.5105 Boiling Point: 120° UN No.: 3399 MDL No.: MFCD00013478
Formula: C₆H₈Si
MW: 108.22 g/mol
Boiling Pt: 120 °C
Density: 0.88
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013478
CAS Number: 694-53-1
EINECS: 211-772-5
UN: 3399
ADR: 4.3 II