Phenylsilane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Silylbenzene
M-Clarity™ quality level = MQ100
335150-1G 335150-25G 335150-5G
SIAL335150-1GEA 30.8 EUR
SIAL335150-1G SIAL335150-25G SIAL335150-5G
Phenylsilane, Sigma-Aldrich®
Phenylsilane
Formula: C₆H₈Si
MW: 108.22 g/mol
Boiling Pt: 118
Density: 0.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013478
CAS Number: 694-53-1
EINECS: 211-772-5
UN: 3399
ADR: 4.3 II