Fentin chloride 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Chlorotriphenylstannane, Triphenyltin chloride
L14289.22 L14289.36
L14289.22EA 42.2 EUR
L14289.22 L14289.36
Fentin chloride 95%
Fentin chloride
Purity: 95% CAS No.: 639-58-7 Molecular Formula: C18H15ClSn Formula Weight: 385.48 Flash Point: 110°(230°F) Odour: Unpleasant Boiling Point: 240°/13.5mm Beilstein Reference: 524762 UN No.: 3146 MDL No.: MFCD00000519
Formula: C₁₈H₁₅ClSn
MW: 385.48 g/mol
Boiling Pt: 240 °C (13.5 mmHg)
Melting Pt: 103…106 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000519
CAS Number: 639-58-7
EINECS: 211-358-4
UN: 3146