Phosphorus(III) bromide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
288462-10G 288462-50G
SIAL288462-10GEA 93.2 EUR
SIAL288462-10G SIAL288462-50G
Phosphorus(III) bromide, Sigma-Aldrich®
Phosphorus (III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –40 °C
Density: 2.8903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514