Antimony(V) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Antimony pentachloride
M-Clarity™ quality level = MQ100
215171-1KG 215171-250G 215171-50G
SIAL215171-1KGEA 363 EUR
SIAL215171-1KG SIAL215171-250G SIAL215171-50G
Antimony(V) chloride, Sigma-Aldrich®
Antimony (V) chloride
Formula: Cl₅Sb
MW: 299.02 g/mol
Boiling Pt: 140
Melting Pt: 3
Density: 2.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8 II
Merck Index: 13,00701