Antimony(V) chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Antimony pentachloride
M-Clarity™ quality level = MQ100
338877-25G
338877-5G
SIAL338877-25GEA
183
EUR
SIAL338877-25G
SIAL338877-5G
Antimony(V) chloride, Sigma-Aldrich®
Antimony (V) chloride
Formula:
Cl₅Sb MW: 299.02 g/mol Boiling Pt: 140 °C (1013 hPa) Melting Pt: 3 °C Density: 2.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011213 CAS Number: 7647-18-9 EINECS: 231-601-8 UN: 1730 ADR: 8,II Merck Index: 13,00701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...