Bismuth(III) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
224839-25G 224839-500G 224839-100G
SIAL224839-25GEA 99.1 EUR
SIAL224839-25G SIAL224839-500G SIAL224839-100G
Bismuth(III) chloride, Sigma-Aldrich®
Bismuth (III) chloride
Formula: BiCl₃
Boiling Pt: 447 °C (1013 hPa)
Melting Pt: 232 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 13,01261