Phosphorus(V) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
157775-1KG 157775-250G
SIAL157775-1KGEA 120 EUR
SIAL157775-1KG SIAL157775-250G
Phosphorus(V) chloride, Sigma-Aldrich®
Phosphorus (V) chloride
Formula: Cl₅P
MW: 208.24 g/mol
Melting Pt: ∼148 °C
Density: 3.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011439
CAS Number: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II
Merck Index: 12,07508