Phosphoryl trichloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride

M-Clarity™ quality level = MQ200

201170-1KG 201170-250G 201170-5G
SIAL201170-1KGEA 103 EUR
SIAL201170-1KG SIAL201170-250G SIAL201170-5G
Phosphoryl trichloride, Sigma-Aldrich®
Phosphoryl trichloride
Formula: POCl₃
MW: 153.33 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 8,I
Merck Index: 12,07506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR