Bismuth(III) nitrate pentahydrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL254150-25GEA 123 EUR
NOVS254150-25G SIAL254150-25G
Bismuth(III) nitrate pentahydrate, Sigma-Aldrich®
Bismuth(III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485,07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C
Density: 2,736 g/cm³ (20 °C)
MDL Number: MFCD00149157
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR