Bismuth(III) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

M-Clarity™ quality level = MQ200

223891-100G 223891-500G
SIAL223891-100GEA 75.8 EUR
SIAL223891-100G SIAL223891-500G
Bismuth(III) oxide, Sigma-Aldrich®
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890 °C (1013 hPa)
Melting Pt: 825 °C
Density: 8.9 g/cm³ (20 °C)
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR