Bismuth(III) oxide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
M-Clarity™ quality level = MQ200
95381-100G
95381-500G
SIAL95381-100GEA
69.9
EUR
SIAL95381-100G
SIAL95381-500G
Bismuth(III) oxide, Sigma-Aldrich®
Bismuth (III) oxide
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: 1890 °C (1013 hPa) Melting Pt: 825 °C Density: 8.9 g/cm³ (20 °C) |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...