Diphosphorus pentoxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL214701-100GEA 17.7 EUR
SIAL214701-100G SIAL214701-500G SIAL214701-1KG SIAL214701-2.5KG
Diphosphorus pentoxide, Sigma-Aldrich®
Diphosphorus pentoxide
Formula: P₂O₅
MW: 141,94 g/mol
Melting Pt: 562 °C
Density: 2,253 g/cm³ (20 °C)
MDL Number: MFCD00011440
UN: 1807
ADR: 8,II
Merck Index: 13,07441