Tributyl phosphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TBP, TBPA
M-Clarity™ quality level = MQ200
90820-100ML 90820-500ML
SIAL90820-100MLEA 66.6 EUR
SIAL90820-100ML SIAL90820-500ML
Tributyl phosphate, Sigma-Aldrich®
Tributyl phosphate
Formula: (CH₃(CH₂)₃O)₃PO
MW: 266.32 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.982 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM