Triethyl phosphite, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
T61204-100ML T61204-1L T61204-500ML T61204-5ML
SIALT61204-100MLEA 26.4 EUR
SIALT61204-100ML SIALT61204-1L SIALT61204-500ML SIALT61204-5ML
Triethyl phosphite, Sigma-Aldrich®
Triethyl phosphite
Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: 51 °C
Density: 0.9643 g/cm³ (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009084
CAS Number: 122-52-1
EINECS: 204-552-5
UN: 2323
ADR: 3,III