Trimethyl phosphite, Sigma-Aldrich®
Supplier: Merck
Synonyms: TMP
M-Clarity™ quality level = MQ200
92730-1L 92730-250ML 92730-50ML
SIAL92730-1LEA 120 EUR
SIAL92730-1L SIAL92730-250ML SIAL92730-50ML
Trimethyl phosphite, Sigma-Aldrich®
Trimethyl phosphite
Formula: C₃H₉O₃P
MW: 124.08 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –77.8 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00008350
CAS Number: 121-45-9
EINECS: 204-471-5
UN: 2329
ADR: 3,III