Antimony (III) sulphide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black

M-Clarity™ quality level = MQ100

229466-50G 229466-5G
SIAL229466-50GEA 827 EUR
SIAL229466-50G SIAL229466-5G
Antimony (III) sulphide, Sigma-Aldrich®
Antimony (III) sulphide
Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150
Melting Pt: 550
Density: 4.12 g/cm³ (20 °C)
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1 III
Merck Index: 12,00754

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR