Antimony (III) sulphide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black
M-Clarity™ quality level = MQ100
244562-100G 244562-500G
SIAL244562-100GEA 45.9 EUR
SIAL244562-100G SIAL244562-500G
Antimony (III) sulphide, Sigma-Aldrich®
Antimony (III) sulphide
Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150
Melting Pt: 550
Density: 4.12 g/cm³ (20 °C)
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1 III
Merck Index: 12,00754